Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MPGFLVRILL LLLVLLLLGP TRGLRNATQR MFEIDYSRDS FLKDGQPFRY ISGSIHYSRV PRFYWKDRLL KMKMAGLNAI QTYVPWNFHE PWPGQYQFSE DHDVEYFLRL AHELGLLVIL RPGPYICAEW EMGGLPAWLL EKESILLRSS DPDYLAAVDK WLGVLLPKMK PLLYQNGGPV ITVQVENEYG SYFACDFDYL RFLQKRFRHH LGDDVVLFTT DGAHKTFLKC GALQGLYTTV DFGTGSNITD AFLSQRKCEP KGPLINSEFY TGWLDHWGQP HSTIKTEAVA SSLYDILARG ASVNLYMFIG GTNFAYWNGA NSPYAAQPTS YDYDAPLSEA GDLTEKYFAL RNIIQKFEKV PEGPIPPSTP KFAYGKVTLE KLKTVGAALD ILCPSGPIKS LYPLTFIQVK QHYGFVLYRT TLPQDCSNPA PLSSPLNGVH DRAYVAVDGI PQGVLERNNV ITLNITGKAG ATLDLLVENM GRVNYGAYIN DFKGLVSNLT LSSNILTDWT IFPLDTEDAV RSHLGGWGHR DSGHHDEAWA HNSSNYTLPA FYMGNFSIPS GIPDLPQDTF IQFPGWTKGQ VWINGFNLGR YWPARGPQLT LFVPQHILMT SAPNTITVLE LEWAPCSSDD PELCAVTFVD RPVIGSSVTY DHPSKPVEKR LMPPPPQKNK DSWLDHV
Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid beta-galactosidase; Beta-Gal; Beta-galactosidase; betagal; bgal; Elastin receptor 1; elastin receptor 1, 67kDa; Lactase; ß-gal; ß-galactosidase; ßgal
Gene Aliases: EBP; ELNR1; GLB1; MPS4B
UniProt ID: (Human) P16278
Entrez Gene ID: (Human) 2720
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support