Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RRLDHIVMTV KSIKDTTMFY SKILGMEVMT FKEDRKALCF GDQKFNLHEV GKEFEPKAAH PVPGSLDICL ITEVPLEEMI QHLKACDVPI EEGPVPRTGA KGPIMSIYFR DPDRNLIEVS NY
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glyoxalase domain-containing protein 5
Gene Aliases: GLOD5
UniProt ID: (Human) A6NK44
Entrez Gene ID: (Human) 392465
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support