Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • GLUT3 Antibodies

          Alomone Labs, LTD

          GLUT3 (extracellular) Polyclonal Antibody

          View all (21) GLUT3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite GLUT3 (extracellular) Polyclonal Antibody

          • Antibody Testing Data (3)
          GLUT3 (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          GLUT3 (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GLUT3 (extracellular) Antibody (AGT-023-200UL) in IHC (F)

          Expression of Glucose transporter 3 in mouse hippocampus and cerebellum - Immunohistochemical staining of perfusion-fixed frozen mouse brain sections with Anti-GLUT3 (extracellular) Antibody (#AGT-023), (1:200), followed by goat Anti-rabbit-AlexaFluor-488. A. GLUT3 staining (green) in mouse hippocampal dentate gyrus, is detected in interneurons (arrows pointing up) in the Hilus and granule layer (G) (arrow ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GLUT3 (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GLUT3 (extracellular) Antibody in Western Blot (WB)
          GLUT3 (extracellular) Antibody in Flow Cytometry (Flow)

          Product Details

          AGT-023-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:400
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:200
          -

          Flow Cytometry (Flow)

          5 µg
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          (C)KDFLNYTLEERLED, corresponding to amino acid residues 39-52 of rat Glucose Transporter 3, 1st extracellular loop
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.8 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).

          Target Information

          Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF1 induced signaling. GLUT3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT3 staining is also detectable in human cancers including those of the ovary, lung, and testis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ90380; glucose transporter 3; Glucose transporter type 3, brain; GLUT-3; Solute carrier family 2 A3 (neuron glucose transporter); Solute carrier family 2, facilitated glucose transporter member 3; solute carrier family 2, member 2; solute carrier family 2, member 3

          View more View less

          Gene Aliases: AA408729; AL023014; AL024341; AU040424; C78366; Glut-3; GLUT3; Slc2a3

          View more View less

          UniProt ID: (Mouse) P32037, (Rat) Q07647

          View more View less

          Entrez Gene ID: (Mouse) 20527, (Rat) 25551

          View more View less

          Function(s)
          transporter activity sugar:proton symporter activity glucose transmembrane transporter activity glucose binding monosaccharide transmembrane transporter activity galactoside binding kinase binding transmembrane transporter activity substrate-specific transmembrane transporter activity xylose binding dehydroascorbic acid transporter activity D-glucose transmembrane transporter activity protein binding
          Process(es)
          transport carbohydrate transport glucose transport proton transport hexose transmembrane transport glucose import transmembrane transport dehydroascorbic acid transport cation transmembrane transport response to glucose monosaccharide transport negative regulation of apoptotic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0