Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • GOT2 Antibodies

          Proteintech

          GOT2 Monoclonal Antibody (1B6C6), CoraLite® Plus 488

          View all (23) GOT2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite GOT2 Monoclonal Antibody (1B6C6), CoraLite® Plus 488

          • Antibody Testing Data (1)
          GOT2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          GOT2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GOT2 Antibody (CL488-67738) in Flow

          1X10^6 HeLa cells were intracellularly stained with 0.4 µg CoraLite® Plus 488 Anti-Human GOT2 (CL488-67738, Clone:1B6C6) (red), or 0.4 µg Control Antibody. Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GOT2 Antibody in Flow Cytometry (Flow)

          Product Details

          CL488-67738

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.4 µg/1x10^6 cell suspension
          -
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse, Pig, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1B6C6

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Purification

          Protein G

          Storage buffer

          PBS with 0.5% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: KAEAQIAAKN LDKEYLPIGG LAEFCKASAE LALGENSEVL KSGRFVTVQT ISGTGALRIG ASFLQRFFKF SRDVFLPKPT WGNHTPIFRD AGMQLQGYRY YDPKTCGFDF TGAVEDISKI PEQSVLLLHA CAHNPTGVDP RPEQWKEIAT VVKKRNLFAF FDMAYQGFAS GDGDKDAWAV RHFIEQGINV CLCQSYAKNM GLYGERVGAF TMVCKDADEA KRVESQLKIL IRPMYSNPPL NGARIAAAIL NTPDLRKQWL QEVKVMADRI IGMRTQLVSN LKKEGSTHNW QHITDQIGMF CFTGLKPEQV ERLIKEFSIY MTKDGRISVA GVTSSNVGYL AHAIHQATK

          Target Information

          GOT2 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: aspartate aminotransferase 2; Aspartate aminotransferase, mitochondrial; aspartate transaminase 2; FABP-1; FABPpm; Fatty acid-binding protein; FLJ40994; Glutamate oxaloacetate transaminase 2; glutamate oxaloacetate transaminase 2, mitochondrial; Glutamate oxaloacetate transaminase 2, mitochondrial (aspartate aminotransferase 2); glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase 2); Kynurenine aminotransferase 4; Kynurenine aminotransferase IV; Kynurenine--oxoglutarate transaminase 4; Kynurenine--oxoglutarate transaminase IV; mAspAT; mitochondrial aspartate aminotransferase; plasma membrane fatty acid binding protein; Plasma membrane-associated fatty acid-binding protein; Transaminase A

          View more View less

          Gene Aliases: AL022787; ASPATA; FABP-pm; Got-2; GOT2; KAT4; KATIV; KYAT4; Maat; mAspAT; mitAAT

          View more View less

          UniProt ID: (Human) P00505, (Pig) P00506, (Chicken) P00508, (Mouse) P05202, (Rat) P00507

          View more View less

          Entrez Gene ID: (Human) 2806, (Pig) 396968, (Chicken) 396533, (Mouse) 14719, (Rat) 25721

          View more View less

          Function(s)
          L-aspartate:2-oxoglutarate aminotransferase activity phospholipid binding kynurenine-oxoglutarate transaminase activity amino acid binding enzyme binding pyridoxal phosphate binding identical protein binding protein homodimerization activity poly(A) RNA binding L-phenylalanine:2-oxoglutarate aminotransferase activity catalytic activity transaminase activity transferase activity carboxylic acid binding
          Process(es)
          oxaloacetate metabolic process cellular amino acid metabolic process aspartate metabolic process aspartate biosynthetic process aspartate catabolic process transport lipid transport biosynthetic process glutamate catabolic process to aspartate glutamate catabolic process to 2-oxoglutarate dicarboxylic acid metabolic process response to ethanol gluconeogenesis 2-oxoglutarate metabolic process glutamate metabolic process cellular amino acid biosynthetic process fatty acid transport 4-hydroxyproline catabolic process L-kynurenine metabolic process carbohydrate metabolic process small molecule metabolic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0