Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This target displays homology in the following species: Human: 100%
GPAT2 gene ontology annotations related to this gene include cellular lipid metabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase GPAT2; cancer/testis antigen 123; Glycerol-3-phosphate acyltransferase 2, mitochondrial; GPAT-2; xGPAT1
Gene Aliases: CT123; GPAT2
UniProt ID: (Human) Q6P2E4
Entrez Gene ID: (Human) 150763
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support