Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PGLPMLRQIR EARRKEEKQQ EANLKNRQKS LKEEEQERRD IGLKNALGCE NKGFALLQKM GYKSGQALGK SGGGIVEPIP LNIKT
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centromere protein Y; coiled-coil domain containing 75; Coiled-coil domain-containing protein 75; G patch domain containing 11; G patch domain-containing protein 11
Gene Aliases: CCDC75; CENP-Y; CENPY; GPATCH11
UniProt ID: (Human) Q8N954
Entrez Gene ID: (Human) 253635
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support