Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: NDEGRQGDDE QSDWFYEKES GGACGITGVV PWWEKEDPTE LDKNVPDPVF ESILTGSFPL MSHPSRRGFQ ARLSRLHGMS SKNIKKSGGT PTSMVPIPGP V
Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 75%.
The gene encodes a nuclear factor that may play a role in spermatogenesis and in tumor growth during breast cancer. The encoded protein contains a G-patch domain with an RNA binding motif. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 110; G patch domain containing 2; G patch domain-containing protein 2; protein phosphatase 1, regulatory subunit 30; RP11-361K17.1
Gene Aliases: CT110; GPATC2; GPATCH2; PPP1R30
UniProt ID: (Human) Q9NW75
Entrez Gene ID: (Human) 55105
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support