Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MRTDKKSEF LKALKRDRVE EEHEDESRAG SEKDDDSFNL HNSNSTHQER DINRNFDENE IPQENGNASV ISQQIIRSST FPQTDVLSSS LEAEHRLLKE MGWQEDSEND ETCAPLTEDE MREFQVISEQ LQKNGLRKNG ILKNGLICDF KFGPWKNSTF KPTTENDDTE TSSSDTSDDD DV (293-473 aa encoded by BC000267)
Vasculin is a novel vascular protein differentially expressed in human atherogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP761C169; GC-rich promoter-binding protein 1; mGPBP; Vascular wall-linked protein; Vasculin
Gene Aliases: 1700034P14Rik; AU019836; D230035M11Rik; GPBP; GPBP1; mGPBP; RGD1305492; SSH6; VASCULIN
UniProt ID: (Human) Q86WP2, (Mouse) Q6NXH3
Entrez Gene ID: (Human) 65056, (Mouse) 73274, (Rat) 294734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support