Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR3 is an orphan receptor with constitutive G signaling activity. GPR3 modulates a number of brain functions, including behavioral responses to stress, neurite outgrowth and amyloid-beta peptide generation suggesting that it may play a role in Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor R4; G-protein coupled receptor 3; G-protein-coupled receptor R4; GPCR21; Hacca; OTTHUMP00000043209
Gene Aliases: Gpcr20; Gpcr21; Gpcr3; Gpr3
UniProt ID: (Mouse) P35413, (Rat) Q8K1Q3
Entrez Gene ID: (Mouse) 14748, (Rat) 266769
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support