Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GPRs are characterized by the presence of 7 transmembrane domains. GPR128 is an orphan receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G-protein coupled receptor G7; FLJ14454; FLJ16382; FLJ29035; G protein-coupled receptor 128; G-protein coupled receptor 128; MGC142011; MGC163260
Gene Aliases: ADGRG7; GPR128
UniProt ID: (Human) Q96K78
Entrez Gene ID: (Human) 84873
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support