Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total GPR137C.
GPR137C is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 137C; G protein-coupled receptor TM7SF1L2; Integral membrane protein GPR137C; Transmembrane 7 superfamily member 1-like 2 protein
Gene Aliases: 6330416L11Rik; Gm908; GPR137C; TM7SF1L2
UniProt ID: (Human) Q8N3F9, (Mouse) E9Q343
Entrez Gene ID: (Human) 283554, (Mouse) 70713
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support