Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: ERYRADVRT VWEQCVAIMS EEDGDDDGGC DDYAEGRVCK VRFDANGATG PGSRDPAQVK LLPGRHMLFP PLERVHYLQV PLSRRLSHDE TNIFSTPREP GSFLHKWSSS DDIRVLPAQS RALGGPPEYL GQRHRLEDEE DEEEAEGGGL ASLRQFLESG VLGSGGGPPR GPGFFREEIT TFIDETPLPS PTASPGHSPR RPRPLGLSPR RLSLGSPESR AVGLPLGLSA GRRCSLTGGE ESARAWGGSW GPGNPIFPQL TL (328-588 aa encoded by BC002353)
This gene was identified upon genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be mainly expressed in the brain; however, its function is not known. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene rich cluster, A; Gene-rich cluster gene A protein; Probable G-protein coupled receptor 162
Gene Aliases: A-2; GPR162; GRCA
UniProt ID: (Human) Q16538
Entrez Gene ID: (Human) 27239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support