Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VPYHVNRSV YVLHYRSHGA SCATQRILAL ANRITSCLTS LNGALDPIMY FFVAEKFRHA LCNLLCGKRL KGPPPSFEGK TNESSLSAKS EL (283-367 aa encoded by BC031653)
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR17 is an orphan G protein-coupled receptor involved in orchestration of oligodendrocyte differentiation and myelination in the central nervous system. GPR17 is expressed in brain, kidney, heart and umbilical vein endothelial cells. GPR17 has dual specificity for uracil nucleotides and cysteinyl leukotrienes and signals through G(i) and inhibition of adenylyl cyclase. May mediate brain damage by nucleotides and cysteinyl leukotrienes following ischemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686M18273; G-protein coupled receptor 17; gpr17; gpr17 {ECO:0000312|RGD:1589785}; P2Y-like receptor; R12; UDP/CysLT receptor; Uracil nucleotide/cysteinyl leukotriene receptor; Y12546; Z94155
Gene Aliases: AI853548; GPR17
UniProt ID: (Human) Q13304, (Mouse) Q6NS65, (Rat) Q09QM4
Entrez Gene ID: (Human) 2840, (Mouse) 574402, (Rat) 767613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support