Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein that is a member of the G protein-coupled receptor superfamily. This protein is produced predominantly in vascular smooth muscle cells and may play an important role in the regulation of vascular remodeling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Integral membrane protein GPR180; Intimal thickness-related receptor
Gene Aliases: AB041545; E130016I23Rik; GPR180; ITR; MNCb-3029
UniProt ID: (Human) Q86V85, (Mouse) Q8BPS4
Entrez Gene ID: (Human) 160897, (Rat) 306165, (Mouse) 58245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support