Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • GPR30 Antibodies

          Bioss

          GPR30 Polyclonal Antibody, Alexa Fluor™ 594

          View all (45) GPR30 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite GPR30 Polyclonal Antibody, Alexa Fluor™ 594

          Product Details

          BS-20643R-A594

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:20-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human GPR30
          View immunogen

          Conjugate

          Alexa Fluor™ 594 Alexa Fluor™ 594 Alexa Fluor™ 594

          Excitation/Emission Max

          590/618 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.03% ProClin 300

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          This gene is a member of the G-protein coupled receptor 1 family and encodes a multi-pass membrane protein that localizes to the endoplasmic reticulum. The protein binds estrogen, resulting in intracellular calcium mobilization and synthesis of phosphatidylinositol 3,4,5-trisphosphate in the nucleus. This protein therefore plays a role in the rapid nongenomic signaling events widely observed following stimulation of cells and tissues with estrogen. Alternate transcriptional splice variants which encode the same protein have been characterized.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Chemoattractant receptor-like 2; chemokine receptor-like 2; constitutively expressed peptide-like receptor; constitutively expressed peptide-like receptor like; FEG-1; Flow-induced endothelial G-protein coupled receptor 1; G protein-coupled estrogen receptor 1; G protein-coupled receptor 30; G-protein coupled estrogen receptor 1; G-protein coupled receptor 30; G-protein coupled receptor 41; GPCR-Br; heptahelix receptor; IL8-related receptor DRY12; leucine rich protein in GPR30 3'UTR; LYGPR; Lymphocyte-derived G-protein coupled receptor; Membrane estrogen receptor; mER

          View more View less

          Gene Aliases: 6330420K13Rik; CEPR; Ceprl; CMKRL2; DRY12; FEG-1; GPCR-Br; GPER; GPER1; GPR30; GPR41; LERGU; LERGU2; LyGPR; mER

          View more View less

          UniProt ID: (Human) Q99527, (Rat) O08878, (Mouse) Q8BMP4

          View more View less

          Entrez Gene ID: (Human) 2852, (Rat) 171104, (Mouse) 76854

          View more View less

          Function(s)
          chromatin binding G-protein coupled receptor activity steroid binding protein binding drug binding mineralocorticoid receptor activity estrogen receptor activity steroid hormone binding steroid hormone receptor activity hormone binding signal transducer activity
          Process(es)
          positive regulation of protein phosphorylation positive regulation of neurotransmitter secretion negative regulation of leukocyte activation inflammatory response positive regulation of cytosolic calcium ion concentration positive regulation of cell proliferation negative regulation of cell proliferation positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway positive regulation of gene expression negative regulation of gene expression positive regulation of phosphatidylinositol 3-kinase signaling neuronal action potential apoptotic chromosome condensation nuclear fragmentation involved in apoptotic nuclear change positive regulation of cell migration intracellular steroid hormone receptor signaling pathway positive regulation of cAMP biosynthetic process mineralocorticoid receptor signaling pathway positive regulation of insulin secretion positive regulation of inositol trisphosphate biosynthetic process positive regulation of apoptotic process positive regulation of cysteine-type endopeptidase activity involved in apoptotic process steroid hormone mediated signaling pathway positive regulation of MAPK cascade innate immune response negative regulation of fat cell differentiation positive regulation of epidermal growth factor receptor signaling pathway positive regulation of G-protein coupled receptor protein signaling pathway positive regulation of vasodilation positive regulation of transcription from RNA polymerase II promoter negative regulation of inflammatory response positive regulation of neurogenesis negative regulation of DNA metabolic process negative regulation of lipid biosynthetic process positive regulation of release of sequestered calcium ion into cytosol regulation of cytosolic calcium ion concentration positive regulation of ERK1 and ERK2 cascade positive regulation of uterine smooth muscle contraction negative regulation of cell cycle arrest cellular response to glucose stimulus cellular response to tumor necrosis factor cellular response to peptide hormone stimulus cellular response to mineralocorticoid stimulus cellular response to estradiol stimulus positive regulation of establishment of protein localization to plasma membrane positive regulation of release of cytochrome c from mitochondria positive regulation of endothelial cell apoptotic process positive regulation of extrinsic apoptotic signaling pathway cell cycle G-protein coupled receptor signaling pathway negative regulation of cell cycle process negative regulation of protein kinase B signaling negative regulation of ERK1 and ERK2 cascade negative regulation of vascular smooth muscle cell proliferation positive regulation of cardiac vascular smooth muscle cell differentiation immune system process transcription from RNA polymerase II promoter apoptotic process signal transduction nervous system development cell differentiation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0