Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive Control: and spinal chord at low to moderate levels. colon Expressed at high level in fetal and adult lung tissues. Constitutively expressed in fetal kidney and adult placenta kidney ovary prostate Small Intestine stomach testis
Cellular Location: Cell membrane. Cytoplasmic vesicle membrane. Localized in the plasma membrane and perinuclear vesicles.
Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function for this retinoic acid-induced gene has not yet been determined; however, it may play a role in embryonic development and epithelial cell differentiation. This gene is a member of the type 3 G protein-coupling receptor family, characterized by the signature 7-transmembrane domain motif. This gene may also be involved in interaction between retinoid acid and G protein signalling pathways. RAI3 has been reported in lung, kidney, colon, pancreas, prostate, and small intestine. ESTs have been isolated from amnion, breast, colon, eye, lung, nasal cavity, placenta, skin, uterus, and vessel libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor, class C, group 5, member A; G protein-coupled receptor, family C, group 5, member A; G-protein coupled receptor family C group 5 member A; orphan G-protein-coupling receptor PEIG-1; PEIG-1; Phorbol ester induced gene 1; phorbol ester induced protein-1; RAIG-1; retinoic acid induced 3; retinoic acid responsive; Retinoic acid-induced gene 1 protein; Retinoic acid-induced protein 3; TPA induced gene 1
Gene Aliases: GPCR5A; GPRC5A; PEIG-1; RAI3; RAIG1; TIG1
UniProt ID: (Human) Q8NFJ5
Entrez Gene ID: (Human) 9052
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support