Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Predicted to react in Rat
GRAMD4 also termed as Death inducing protein is a 578 amino-acid protein, which contains 1 GRAM domain. GRAMD4 as a multi-pass membrane protein localizes in the Mitochondrion membrane. Overexpression of GRAMD4 results in a strong apoptotic response involving caspase-3 activation and cleavage of poly (ADP-ribose)-polymerase. GRAMD4 is expressed in lung and in primary lung squamous cell carcinoma (LSCC) and shows up-regulation in mitochondria by E2F-1 after addition of 4-hydroxytamoxifen. This evidence suggests that GRAMD4 may be a potential target for cancer therapies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Death-inducing protein; death-inducing-protein; GRAM domain-containing protein 4; GRAM domain-containing protein 4 (GRAMD4)
Gene Aliases: dA59H18.1; DIP; dJ439F8.1; GRAMD4; KIAA0767
UniProt ID: (Human) Q6IC98
Entrez Gene ID: (Human) 23151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support