Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 13
Immunogen sequence: SGELLSSLPP TSNAPSDPAT TTAKADAASS LTVDVTPPTA KAPTTVEDRV GDSTPVSEKP VSAAVDANAS ESP
Binds to an epitope located within the peptide sequence DAASSLTVDVTPPTA as determined by overlapping synthetic peptides.
GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Golgi phosphoprotein 6; golgi reassembly stacking protein 2, 55kDa; Golgi reassembly-stacking protein 2; Golgi reassembly-stacking protein of 55 kDa; GOLPH6; GRASP55; GRS2; p59
Gene Aliases: GOLPH6; GORASP2; GRASP55; GRS2; p59
UniProt ID: (Human) Q9H8Y8
Entrez Gene ID: (Human) 26003
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support