Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MELENIVANT VLLKAREGGG GNRKGKSKKW RQMLQFPHIS QCEELRLSLE RDYHSLCERQ PIGRLLFREF CATRPELSRC VAFLDGVAEY EVTPDDKRKA CGRQLTQNFL SHTGPDLIPE VPRQLVTNCT QRLEQGPCKD LFQELTRLTH EYLSVAPFAD YLDSIYFNRF LQWKWLERQP VTKNTFRQYR VLGKGGFGEV CACQVRATGK MYACKKLEKK RIKKRKGEAM ALNEKQILEK VNSRFVVSLA YAYETKDALC LVLTLMNGGD LKFHIYHMGQ AGFPEARAVF YAAEICCGLE DLHRERIVYR DLKPENILLD DHGHIRISDL GLAVHVPEGQ TIKGRVGTVG YMAPEVVKNE RYTFSPDWWA LGCFLYEMIA GQSPFQQRKK KIKREEVERL VKEVPEEYSE RFSPQARSLC SQLLCKDPAE RLGCRGGSAR EVKEHPLFKK LNFKRLGAGM LEPPFKPDPQ AIYCKDVLDI EQFSTVKGVE LEPTDQDFYQ KFATGSVPIP WQNEMVETEC FQELNVFGLD GSVPPDLDWK GQPPAPPKKG LLQRLFSRQR IAVETAATAR KSSPPASSPQ PEAPTSSWR
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation. Several transcript variants encoding different isoforms have been described for GRK6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.11.16; G protein-coupled receptor kinase 6; G protein-coupled receptor kinase GRK6; kinase GRK6
Gene Aliases: GPRK6; GRK6
UniProt ID: (Human) P43250
Entrez Gene ID: (Human) 2870
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support