Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MGSAFERVVR RVVQELDHGG EFIPVTSLQS STGFQPYCLV VRKPSSSWFW KPRYKCVNLS IKDILEPDAA EPDVQRGRSF HFYDAMDGQI QGSVELAAPG QAKIAGGAAV SDSSSTSMNV YSLSVDPNTW QTLLHERHLR QPEHKVLQQL RSRGDNVYVV TEVLQTQKEV EVTRTHKREG SGRFSLPGAT CLQGEGQGHL SQKKTVTIPS GSTLAFRVAQ LVIDSDLDVL LFPDKKQRTF QPPATGHKRS TSEGAWPQLP SGLSMMRCLH NFLTDGVPAE GAFTEDFQGL RAEVETISKE LELLDRELCQ LLLEGLEGVL RDQLALRALE EALEQGQSLG PVEPLDGPAG AVLECLVLSS GMLVPELAIP VVYLLGALTM LSETQHKLLA EALESQTLLG PLELVGSLLE QSAPWQERST MSLPPGLLGN SWGEGAPAWV LLDECGLELG EDTPHVCWEP QAQGRMCALY ASLALLSGLS QEPH
GSDMD (gasdermin-D) promotes pyroptosis in response to microbial infection and danger signals. It is produced by the cleavage of gasdermin-D by inflammatory caspases CASP1 or CASP4 in response to canonical, as well as, non-canonical (such as cytosolic LPS) inflammasome activators. After cleavage, GSDMD moves to the plasma membrane where it strongly binds to inner leaflet lipids including phosphatidylinositol 4-phosphate, phosphatidylinositol (4,5)-biphosphate, phosphatidylinositol (3,4,5)-biphosphate, phosphatidic acid, and phosphatidylserine. GSDMD also exhibits bactericidal activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gasdermin domain containing 1; Gasdermin domain-containing protein 1; Gasdermin-D
Gene Aliases: DF5L; DFNA5L; FKSG10; GSDMD; GSDMDC1
UniProt ID: (Human) P57764
Entrez Gene ID: (Human) 79792
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support