Product References
Molecular Dynamics Simulations Reveal Novel Interacting Regions of Human Prion Protein to Brucella abortus Hsp60 Protein.
Molecular biotechnology
Le-Dao HA,Dinh TT,Tran TL,Lee VS,Tran-Van H
Mon Apr 01 00:00:00 UTC 2024
Phosphoglycerate dehydrogenase activates PKM2 to phosphorylate histone H3T11 and attenuate cellular senescence.
Nature communications
Wu Y,Tang L,Huang H,Yu Q,Hu B,Wang G,Ge F,Yin T,Li S,Yu X
Fri Mar 10 00:00:00 UTC 2023
Jasmonate activates a CsMPK6-CsMYC2 module that regulates the expression of β-citraurin biosynthetic genes and fruit coloration in orange (Citrus sinensis).
The Plant cell
Yue P,Jiang Z,Sun Q,Wei R,Yin Y,Xie Z,Larkin RM,Ye J,Chai L,Deng X
Wed Mar 29 00:00:00 UTC 2023
SUE4, a novel PIN1-interacting membrane protein, regulates acropetal auxin transport in response to sulfur deficiency.
The New phytologist
Zhao Q,Zhao PX,Wu Y,Zhong CQ,Liao H,Li CY,Fu XD,Fang P,Xu P,Xiang CB
Sun Jan 01 00:00:00 UTC 2023
TRIM28 represses renal cell carcinoma cell proliferation by inhibiting TFE3/KDM6A-regulated autophagy.
The Journal of biological chemistry
Song T,Lv S,Ma X,Zhao X,Fan L,Zou Q,Li N,Yan Y,Zhang W,Sun L
Mon May 01 00:00:00 UTC 2023
PEBP balances apoptosis and autophagy in whitefly upon arbovirus infection.
Nature communications
Wang S,Guo H,Zhu-Salzman K,Ge F,Sun Y
Fri Feb 11 00:00:00 UTC 2022
HERC3 directly targets RPL23A for ubiquitination degradation and further regulates Colorectal Cancer proliferation and the cell cycle.
International journal of biological sciences
Zhang Z,Wu Q,Fang M,Liu Y,Jiang J,Feng Q,Hu R,Xu J
Thu Jun 02 00:00:00 UTC 2022
A novel NF2 splicing mutant causes neurofibromatosis type 2 via liquid-liquid phase separation with large tumor suppressor and Hippo pathway.
iScience
Jia Z,Yang S,Li M,Lei Z,Ding X,Fan M,Wang D,Xie D,Zhou H,Qiu Y,Zhuang Q,Li D,Yang W,Qi X,Cang X,Zhao JW,Wang W,Lin A,Yan Q
Fri Nov 18 00:00:00 UTC 2022
Ubiquitination and degradation of MGMT by TRIM72 increases the sensitivity of uveal melanoma cells to Dacarbazine treatment.
Cancer biomarkers : section A of Disease markers
Li X,Yang C,Luo N,Yang Y,Guo Y,Chen P,Cun B
Thu Jun 02 00:00:00 UTC 2022
Lysine demethylase KDM1A promotes cell growth via FKBP8-BCL2 axis in hepatocellular carcinoma.
The Journal of biological chemistry
Lv S,Zhao X,Zhang E,Yan Y,Ma X,Li N,Zou Q,Sun L,Song T
Thu Sep 01 00:00:00 UTC 2022
HERC3 regulates epithelial-mesenchymal transition by directly ubiquitination degradation EIF5A2 and inhibits metastasis of colorectal cancer.
Cell death & disease
Zhang Z,He G,Lv Y,Liu Y,Niu Z,Feng Q,Hu R,Xu J
Fri Jan 21 00:00:00 UTC 2022
Degradation of lipid droplets by chimeric autophagy-tethering compounds.
Cell research
Fu Y,Chen N,Wang Z,Luo S,Ding Y,Lu B
Wed Sep 01 00:00:00 UTC 2021
Host factor cyclophilin B affects Orf virus replication by interacting with viral ORF058 protein.
Veterinary microbiology
Zhou Y,Gao F,Lv L,Wang S,He W,Lan Y,Li Z,Lu H,Song D,Guan J,Zhao K
Thu Jul 01 00:00:00 UTC 2021
TNPO1-Mediated Nuclear Import of FUBP1 Contributes to Tumor Immune Evasion by Increasing NRP1 Expression in Cervical Cancer.
Journal of immunology research
Yang B,Chen J,Teng Y
Mon Nov 29 00:00:00 UTC 2021
Protein 4.1R negatively regulates P815 cells proliferation by inhibiting C-Kit-mediated signal transduction.
Experimental cell research
Ding C,Guo Y,Liang T,Liu J,Yang L,Wang T,Liu X,Kang Q
Fri Jan 15 00:00:00 UTC 2021
Di-methylation of CD147-K234 Promotes the Progression of NSCLC by Enhancing Lactate Export.
Cell metabolism
Wang K,Huang W,Chen R,Lin P,Zhang T,Ni YF,Li H,Wu J,Sun XX,Geng JJ,Zhu YM,Nan G,Zhang W,Chen X,Zhu P,Bian H,Chen ZN
Tue Jan 05 00:00:00 UTC 2021
Identification of a Shrimp E3 Ubiquitin Ligase TRIM50-Like Involved in Restricting White Spot Syndrome Virus Proliferation by Its Mediated Autophagy and Ubiquitination.
Frontiers in immunology
Zhao C,Peng C,Wang P,Yan L,Fan S,Qiu L
Wed Oct 27 00:00:00 UTC 2021
Identification of a TRIM32 from Penaeus monodon is involved in autophagy and innate immunity during white spot syndrome virus infection.
Developmental and comparative immunology
Peng C,Zhao C,Wang PF,Yan LL,Fan SG,Qiu LH
Fri Oct 01 00:00:00 UTC 2021
The SESAME complex regulates cell senescence through the generation of acetyl-CoA.
Nature metabolism
Chen W,Yu X,Wu Y,Tang J,Yu Q,Lv X,Zha Z,Hu B,Li X,Chen J,Ma L,Workman JL,Li S
Thu Jul 01 00:00:00 UTC 2021
The host cellular protein Ndufaf4 interacts with the vesicular stomatitis virus M protein and affects viral propagation.
Virus genes
Pan W,Shen Z,Wang H,He H
Tue Jun 01 00:00:00 UTC 2021
NIK links inflammation to hepatic steatosis by suppressing PPARα in alcoholic liver disease.
Theranostics
Li Y,Chen M,Zhou Y,Tang C,Zhang W,Zhong Y,Chen Y,Zhou H,Sheng L
Wed Apr 07 00:00:00 UTC 2021
The Shigella Type III Secretion Effector IpaH4.5 Targets NLRP3 to Activate Inflammasome Signaling.
Frontiers in cellular and infection microbiology
Wang X,Sun J,Wan L,Yang X,Lin H,Zhang Y,He X,Zhong H,Guan K,Min M,Sun Z,Yang X,Wang B,Dong M,Wei C
Mon May 17 00:00:00 UTC 2021
A PINCH-1-Smurf1 signaling axis mediates mechano-regulation of BMPR2 and stem cell differentiation.
The Journal of cell biology
Guo L,Wang R,Zhang K,Yuan J,Wang J,Wang X,Ma J,Wu C
Mon Nov 04 00:00:00 UTC 2019
The C7orf43/TRAPPC14 component links the TRAPPII complex to Rabin8 for preciliary vesicle tethering at the mother centriole during ciliogenesis.
The Journal of biological chemistry
Cuenca A,Insinna C,Zhao H,John P,Weiss MA,Lu Q,Walia V,Specht S,Manivannan S,Stauffer J,Peden AA,Westlake CJ
Fri Oct 18 00:00:00 UTC 2019
Allele-selective lowering of mutant HTT protein by HTT-LC3 linker compounds.
Nature
Li Z,Wang C,Wang Z,Zhu C,Li J,Sha T,Ma L,Gao C,Yang Y,Sun Y,Wang J,Sun X,Lu C,Difiglia M,Mei Y,Ding C,Luo S,Dang Y,Ding Y,Fei Y,Lu B
Fri Nov 01 00:00:00 UTC 2019
The matrix protein of vesicular stomatitis virus inhibits host-directed transcription of target genes via interaction with the TFIIH subunit p8.
Veterinary microbiology
Pan W,Song D,He W,Lu H,Lan Y,Tong J,Gao F,Zhao K
Fri Sep 01 00:00:00 UTC 2017
The matrix protein of vesicular stomatitis virus inhibits host-directed transcription of target genes via interaction with the TFIIH subunit p8.
Veterinary microbiology
Pan W,Song D,He W,Lu H,Lan Y,Tong J,Gao F,Zhao K
Fri Sep 01 00:00:00 UTC 2017
EIF3i affects vesicular stomatitis virus growth by interacting with matrix protein.
Veterinary microbiology
Pan W,Song D,He W,Lu H,Lan Y,Li H,Gao F,Zhao K
Fri Dec 01 00:00:00 UTC 2017