Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MANSANTNT VPKLYRSVIE DVINDVRDIF LDDGVDEQVL MELKTLWENK LMQSRAVDGF HSEEQQLLLQ VQQQHQPQQQ QHHHHHHHQQ AQPQQTVPQQ AQTQQVLIPA SQQATAPQVI VPDSKLIQHM NASNMSAAAT AATLALPAGV TPVQQILTNS GQLLQVVRAA NGAQYIFQPQ QSVVLQQQVI PQMQPGGVQA PVIQQVLAPL PGGISPQTGV IIQPQQILFT GNKTQVIPTT VAAPTPAQAQ ITATGQQQPQ AQPAQTQAPL VLQVDGTGDT SSEEDEDEEE DYDDDEEEDK EKDGAEDGQV EEEPLNSEDD VSDEEGQELF D (1-330 aa encoded by BC107156)
Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A) and several general initiation factors. One of these factors is TFIIA, which when purified from HeLa extracts consists of 35-, 19-, and 12-kD subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: general transcription factor 2a, 1; general transcription factor IIA 1; General transcription factor IIA subunit 1; general transcription factor IIa, 1 (37kD and 19kD subunits); general transcription factor IIA, 1, 19/37kDa; glucose regulated protein, 58kD pseudogene; TFIIA alpha, p55, isoform 1; TFIIA alpha/beta; TFIIA alpha/beta subunits; TFIIA large subunit; TFIIA-42; TFIIAL; Transcription initiation factor IIA subunit 1; Transcription initiation factor TFIIA 42 kDa subunit
Gene Aliases: 6330549H03Rik; AA536742; AA959775; AW060250; GTF2A1; Tf2a; TF2A1; TFIIA; TFIIA-42; Tfiia1; TfIIAa/b; TFIIAL
UniProt ID: (Human) P52655, (Mouse) Q99PM3, (Rat) O08949
Entrez Gene ID: (Human) 2957, (Mouse) 83602, (Rat) 83830
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support