Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MANSANTNTV PKLYRSVIED VINDVRDIFL DDGVDEQVLM ELKTLWENKL MQSRAVDGFH SEEQQLLLQV QQQHQPQQQQ HHHHHHHQQA QPQQTVPQQA QTQQVLIPAS QQATAPQVIV PDSKLIQHMN ASNMSAAATA ATLALPAGVT PVQQILTNSG QLLQVVRAAN GAQYIFQPQQ SVVLQQQVIP QMQPGGVQAP VIQQVLAPLP GGISPQTGVI IQPQQILFTG NKTQVIPTTV AAPTPAQAQI TATGQQQPQA QPAQTQAPLV LQVDGTGDTS SEEDEDEEED YDDDEEEDKE KDGAEDGQVE EEPLNSEDDV SDEEGQELFD TENVVVCQYD KIHRSKNKWK FHLKDGIMNL NGRDYIFSKA IGDAEW
Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A) and several general initiation factors. One of these factors is TFIIA, which when purified from HeLa extracts consists of 35-, 19-, and 12-kD subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: general transcription factor IIA 1; General transcription factor IIA subunit 1; general transcription factor IIA, 1, 19/37kDa; glucose regulated protein, 58kD pseudogene; TFIIA alpha, p55, isoform 1; TFIIA alpha/beta subunits; TFIIA-42; TFIIAL; Transcription initiation factor IIA subunit 1; Transcription initiation factor TFIIA 42 kDa subunit
Gene Aliases: GTF2A1; TF2A1; TFIIA; TFIIA-42; TFIIAL
UniProt ID: (Human) P52655
Entrez Gene ID: (Human) 2957
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support