Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MADPDVLTEV PAALKRLAKY VIRGFYGIEH ALALDILIRN SCVKEEDMLE LLKFDRKQLR SVLNNLKGDK FIKCRMRVET AADGKTTRHN YYFINYRTLV NVVKYKLDHM RRRIETDERD STNRASFKCP VCSSTFTDLE ANQLFDPMTG TFRCTFCHTE VEEDESAMPK KDARTLLARF NEQIEPIYAL LRETEDVNLA YEILEPEPTE IPALKQSKDH AATTAGAASL AGGHHREAWA TKGPSYEDLY TQNVVINMDD QEDLHRASLE GKSAKERPIW LRESTVQGAY GSEDMKEGGI DMDAFQEREE GHAGPDDNEE VMRALLIHEK KTSSAMAGSV GAAAPVTAAN GSDSESETSE SDDDSPPRPA AVAVHKREED EEEDDEFEEV ADDPIVMVAG RPFSYSEVSQ RPELVAQMTP EEKEAYIAMG QRMFEDLFE
The general transcription factor TFIIE has an essential role in eukaryotic transcription initiation together with RNA polymerase II and other general factors. Human TFIIE consists of two subunits of relative molecular mass 57, 000 (TFIIE-alpha) and 34, 000 (TFIIE-beta) and joins the preinitiation complex after RNA polymerase II and TFIIF. TFIIE-alpha is necessary for transcription initiation together with TFIIE-beta, and recombinant TFIIE-alpha can fully replace the natural subunit in an in vitro transcription assay. Its sequence contains several interesting structural motifs (leucine repeat, zinc finger and helix-turn-helix) and sequence similarities to bacterial sigma factors that suggest direct involvement in the regulation of transcription initiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: General transcription factor IIE 56 kDa subunit; General transcription factor IIE subunit 1; general transcription factor IIE, polypeptide 1, alpha 56kDa; TFIIE alpha subunit; TFIIE-alpha; Transcription initiation factor IIE subunit alpha
Gene Aliases: FE; GTF2E1; TF2E1; TFIIE-A
UniProt ID: (Human) P29083
Entrez Gene ID: (Human) 2960
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support