Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • GUF1 Antibodies

          Proteintech

          GUF1 Polyclonal Antibody

          View all (4) GUF1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite GUF1 Polyclonal Antibody

          • Antibody Testing Data (21)
          GUF1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GUF1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GUF1 Antibody (17489-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using GUF1 antibody 17489-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GUF1 Antibody in Immunocytochemistry (ICC/IF)
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GUF1 Antibody in Western Blot (WB)
          GUF1 Antibody in Western Blot (WB)
          GUF1 Antibody in Western Blot (WB)
          GUF1 Antibody in Western Blot (WB)
          GUF1 Antibody in Western Blot (WB)
          GUF1 Antibody in Western Blot (WB)
          GUF1 Antibody in Western Blot (WB)
          GUF1 Antibody in Immunoprecipitation (IP)

          Product Details

          17489-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2400
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          GUF1 Fusion Protein Ag11579 (318-669 aa encoded by BC036768)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.14 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: VGYLIAGMK DVTEAQIGDT LCLHKQPVEP LPGFKSAKPM VFAGMYPLDQ SEYNNLKSAI EKLTLNDSSV TVHRDSSLAL GAGWRLGFLG LLHMEVFNQR LEQEYNASVI LTTPTVPYKA VLSSSKLIKE HREKEITIIN PAQFPDKSKV TEYLEPVVLG TIITPDEYTG KIMMLCEARR AVQKNMIFID QNRVMLKYLF PLNEIVVDFY DSLKSLSSGY ASFDYEDAGY QTAELVKMDI LLNGNTVEEL VTVVHKDKAH SIGKAICERL KDSLPRQLFE IAIQAAIGSK IIARETVKAY RKNVLAKCYG GDITRKMKLL KRQAEGKKKL RKIGNVEVPK DAFIKVLKTQ SSK (318-669 aa encoded by BC036768)

          Target Information

          Promotes mitochondrial protein synthesis. May act as a fidelity factor of the translation reaction, by catalyzing a one- codon backward translocation of tRNAs on improperly translocated ribosomes. Binds to mitochondrial ribosomes in a GTP-dependent manner.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EF-4; Elongation factor 4 homolog; GTP-binding protein GUF1 homolog; GTPase GUF1; GTPase of unknown function 1; GUF1 GTPase homolog; Ribosomal back-translocase; Translation factor GUF1, mitochondrial

          View more View less

          Gene Aliases: 4631409J12; AA407526; EF-4; GUF1

          View more View less

          UniProt ID: (Human) Q8N442, (Mouse) Q8C3X4

          View more View less

          Entrez Gene ID: (Human) 60558, (Mouse) 231279

          View more View less

          Function(s)
          GTPase activity GTP binding ribosome binding nucleotide binding hydrolase activity
          Process(es)
          translation positive regulation of translation biological_process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0