Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MSVTDQAFVT LATNDIYCQG ALVLGQSLRR HRLTRKLVVL ITPQVSSLLR VILSKVFDEV IEVNLIDSAD YIHLAFLKRP ELGLTLTKLH CWTLTHYSKC VFLDADTLVL SNVDELFDRG EFSAAPDPGW PDCFNSGVFV FQPSLHTHKL LLQHAMEHGS FDGADQGLLN SFFRNWSTTD IHKHLPFIYN LSSNTMYTYS PAFKQFGSSA KVVHFLGSMK PWNYKYNPQS GSVLEQGSVS SSQHQAAFLH LWWTVYQNNV LPLYKSVQAG EARASPGHTL CRSDVGGPCA DSASGVGEPC ENSTPSAGVP CANSPLGSNQ PAQGLPEPTQ IVDETLSLPE GRRSEDMIAC PETETPAVIT CDPLSQPSPQ PADFTETETI LQPANKVESV SSEETFEPSQ ELPAEALRDP SLQDALEVDL AVSVSQISIE EKVKELSPEE ERRKWEEGRI DYMGKDAFAR IQEKLDRFLQ
GYG2 belongs to the glycogenin family. Glycogenin is a self-glycosylating protein involved in the initiation phase of glycogen biosynthesis. Multiple alternatively spliced transcript variants encoding different isoforms have been identified, including the predominant band around 66kDa and a light band around 45kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycogenin glucosyltransferase; Glycogenin-2; GN-2
Gene Aliases: GN-2; GN2; GYG2
UniProt ID: (Human) O15488
Entrez Gene ID: (Human) 8908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support