Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GBX1 gene ontology annotations related to this gene include adult walking behavior; neuron fate commitment; proprioception; sensory neuron axon guidance; spinal cord motor neuron differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gastrulation and brain-specific homeobox protein 1; gastrulation brain homeo box 1; Homeobox protein GBX-1
Gene Aliases: Gbx-1; GBX1; Huh-17
UniProt ID: (Human) Q14549, (Mouse) P82976
Entrez Gene ID: (Human) 2636, (Rat) 246149, (Mouse) 231044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support