Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Glutamate dehydrogenase (EC 1.4.1.3) catalyzes the reversible oxidative deamination of glutamate to alpha-ketoglutarate using NAD and/or NADP as cofactors. See also GLUD1 (MIM 138130).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis tissue sperm binding protein Li 18mP; GDH 1; GDH 2; glutamate dehydrogenase (NAD(P)+); Glutamate dehydrogenase 1, mitochondrial; Glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; Memory-related gene 2 protein; MRG-2; testicular secretory protein Li 14
Gene Aliases: Ac2-281; AI118167; GDH; Gdh-X; GDH1; GDH2; GLUD; GLUD1; GLUD2; Gludeha; Gludl; GLUDP1; MRG-2
UniProt ID: (Human) P00367, (Human) P49448, (Mouse) P26443, (Rat) P10860
Entrez Gene ID: (Human) 2746, (Human) 2747, (Mouse) 14661, (Rat) 24399
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support