Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Granzyme B Antibodies

          Invitrogen

          Granzyme B Monoclonal Antibody (NGZB), APC, eBioscience™

          21 Published Figures
          27 References
          View all (89) Granzyme B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Granzyme B Monoclonal Antibody (NGZB), APC, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (21)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Granzyme B Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Granzyme B Antibody (17-8898-82) in Flow

          Mouse splenocytes were cultured with plate-bound Anti-Mouse CD3e Functional Grade Purified (Product # 16-0031-82) and Anti-Mouse CD28 Functional Grade Purified (Product # 16-0281-82) for 3 days, then cultured with Protein Transport Inhibitor Cocktail (Product # 00-4980-03) for an additional 5 hours. Cells were surface stained with Anti-Mouse CD8a PE (Product # 12-0081-82) followed by intracellular staining with 0.06 µg of Rat IgG2a K Isotype Control APC (Product # 17-4321-81) (left) or 0.06 µg of Anti-Mouse Granzyme B APC (right). Total vi... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)
          Granzyme B Antibody in Flow Cytometry (Flow)

          Product Details

          17-8898-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 27 publications 27 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          NGZB

          Conjugate

          APC APC APC
          • APC-eFluor 780
          • eFluor 450
          • eFluor 660
          • FITC
          • PE 
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2688068

          Product Specific Information

          Description: This NGZB monoclonal antibody reacts with mouse Granzyme B, which is a member of the granzyme serine protease family. Granzyme B is found in the granules of cytotoxic T cells and NK cells. Granzyme B has also been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved has been identified as mannose 6-phosphate receptor. This receptor functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. This NGZB monoclonal antibody does not cross-react to human Granzyme B nor is staining blocked with GB11, suggesting it recognizes a different epitope.

          Applications Reported: This NGZB antibody has been reported for use in intracellular staining and flow cytometric analysis.

          Applications Tested: This NGZB antibody has been tested by intracellular staining followed by flow cytometric analysis of stimulated mouse splenocytes using the Intracellular Fixation and Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to BestProtocols®: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CCP1; CTLA-1; Cytotoxic cell protease 1; Cytotoxic T lymphocyte associated serine esterase 1; Fragmentin-2; Granzyme B(G,H); GranzymeB; Human lymphocyte protein (Hlp); OTTHUMP00000028189

          View more View less

          Gene Aliases: AI553453; CCP-1/C11; CCP1; Ctla-1; Ctla1; GZB; Gzmb

          View more View less

          UniProt ID: (Mouse) P04187

          View more View less

          Entrez Gene ID: (Mouse) 14939

          View more View less

          Function(s)
          serine-type endopeptidase activity protein binding peptidase activity serine-type peptidase activity hydrolase activity
          Process(es)
          T cell mediated cytotoxicity proteolysis apoptotic process granzyme-mediated apoptotic signaling pathway protein processing cytolysis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0