Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HDAC6 Antibodies

          NeoBiotechnologies

          HDAC6 Monoclonal Antibody (PCRP-HDAC6-1A4)

          View all (54) HDAC6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite HDAC6 Monoclonal Antibody (PCRP-HDAC6-1A4)

          • Antibody Testing Data (4)
          HDAC6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HDAC6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HDAC6 Antibody (10013-MSM1-P0) in ICC/IF

          Immunofluorescence analysis of PFA-fixed HeLa cells. HDAC6 Mouse Monoclonal Antibody (PCRP-HDAC6-1A4) followed by goat anti-mouse IgG-CF488 (green). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HDAC6 Antibody in Immunocytochemistry (ICC/IF)
          HDAC6 Antibody in Flow Cytometry (Flow)
          HDAC6 Antibody in Peptide array (ARRAY)
          HDAC6 Antibody in SDS-PAGE (SDS-PAGE)

          Product Details

          10013-MSM1-P0

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          -

          Flow Cytometry (Flow)

          1-2 µg/10^6 cells
          -

          Immunoprecipitation (IP)

          1-2 µg
          -

          Peptide Array (Array)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          PCRP-HDAC6-1A4

          Immunogen

          Recombinant full-length human HDAC6 protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          200 µg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4, with 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Wet ice

          Target Information

          Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ16239; HD6; HDA6; Histone deacetylase 6; Protein deacetylase HDAC6; protein phosphatase 1, regulatory subunit 90; Tubulin-lysine deacetylase HDAC6

          View more View less

          Gene Aliases: CPBHM; HD6; HDAC6; JM21; KIAA0901; PPP1R90

          View more View less

          UniProt ID: (Human) Q9UBN7

          View more View less

          Entrez Gene ID: (Human) 10013

          View more View less

          Function(s)
          core promoter binding actin binding histone deacetylase activity protein binding beta-catenin binding microtubule binding zinc ion binding enzyme binding polyubiquitin binding ubiquitin protein ligase binding NAD-dependent histone deacetylase activity (H3-K14 specific) histone deacetylase binding tubulin deacetylase activity alpha-tubulin binding tau protein binding beta-tubulin binding misfolded protein binding Hsp90 protein binding dynein complex binding
          Process(es)
          protein polyubiquitination transcription, DNA-templated protein deacetylation misfolded or incompletely synthesized protein catabolic process intracellular protein transport organelle organization negative regulation of microtubule depolymerization response to toxic substance positive regulation of signal transduction response to organic substance regulation of receptor activity regulation of autophagy positive regulation of epithelial cell migration negative regulation of hydrogen peroxide metabolic process positive regulation of receptor biosynthetic process macroautophagy histone deacetylation regulation of protein stability lysosome localization peptidyl-lysine deacetylation cellular response to topologically incorrect protein regulation of gene expression, epigenetic ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway protein complex disassembly negative regulation of protein complex disassembly regulation of fat cell differentiation negative regulation of proteolysis negative regulation of transcription, DNA-templated negative regulation of oxidoreductase activity mitochondrion localization response to misfolded protein regulation of microtubule-based movement regulation of androgen receptor signaling pathway regulation of establishment of protein localization cellular response to hydrogen peroxide aggresome assembly polyubiquitinated misfolded protein transport Hsp90 deacetylation response to growth factor histone H3 deacetylation cellular response to misfolded protein positive regulation of chaperone-mediated protein complex assembly tubulin deacetylation mitophagy in response to mitochondrial depolarization positive regulation of hydrogen peroxide-mediated programmed cell death regulation of mitophagy
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0