Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Antibody detects endogenous levels of total HEPACAM.
HEPACAM is involved in regulating cell motility and cell-matrix interactions. HEPACAM may inhibit cell growth through suppression of cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glialCAM; HEPACAM; Hepatic and glial cell adhesion molecule; Hepatocyte cell adhesion molecule; Protein hepaCAM
Gene Aliases: 2900042E01Rik; GlialCAM; HEPACAM; Hepn1; MLC2A; MLC2B; RGD1306811
UniProt ID: (Human) Q14CZ8, (Mouse) Q640R3
Entrez Gene ID: (Human) 220296, (Mouse) 72927, (Rat) 300517
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support