Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MKRPKEPSG SDGESDGPID VGQEGQLSQM ARPLSTPSSS QMQARKKRRG IIEKRRRDRI NSSLSELRRL VPTAFEKQGS SKLEKAEVLQ MTVDHLKMLH ATGGTGFFDA RALAVDFRSI GFRECLTEVI RYLGVLEGPS SRADPVRIRL LSHLNSYAAE MEPSPTPTGP LAFPAWPWSF FHSCPGLPAL SNQLAILGRV PSPVLPGVSS PAYPIPALRT APLRRATGII LPARRNVLPS RGASSTRRAR PLERPATPVP VAPSSRAARS SHIAPLLQSS SPTPPGPTGS AAYVAVPTPN SSSPGPAGRP AGAMLYHSWV SEITEIGAF (1-328 aa encoded by BC006087 )
This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb33; Class B basic helix-loop-helix protein 33; hairy and enhance of split related 3; Hairy and enhancer of split-related protein 3; Hairy-related transcription factor 3; hairy/enhancer-of-split related with YRPW motif 3; Hairy/enhancer-of-split related with YRPW motif-like protein; hes-related family bHLH transcription factor with YRPW motif-like; HEY-like protein; hHeyL; hHRT3; HRT-3; mHRT3
Gene Aliases: BHLHB33; HESR3; HEY3; HEYL; HRT3
UniProt ID: (Human) Q9NQ87, (Mouse) Q9DBX7
Entrez Gene ID: (Human) 26508, (Mouse) 56198, (Rat) 313575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support