Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Hex Antibodies

          Abnova

          HHEX Monoclonal Antibody (3F11)

          View all (32) Hex antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite HHEX Monoclonal Antibody (3F11)

          • Antibody Testing Data (1)
          HHEX Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HHEX Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HHEX Antibody (H00003087-M01) in WB

          HHEX monoclonal antibody (M01), clone 3F11. Western Blot analysis of HHEX expression in human placenta. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HHEX Antibody in Western Blot (WB)

          Product Details

          H00003087-M01

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3F11

          Immunogen

          HHEX (AAH14336, 1 a.a. approximately 270 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MQYPHPGPAA GAVGVPLYAP TPLLQPAHPT PFYIEDILGR GPAAPTPAPT LPSPNSSFTS LVSPYRTPVY EPTPIHPAFS HHSAAALAAA YGPGGFGGPL YPFPRTVNDY THALLRHDPL GKPLLWSPFL QRPLHKRKGG QVRFSNDQTI ELEKKFETQK YLSPPERKRL TKMLQLSERQ VKTWFQNRRA KWRRLKQENP QSNKKEELES LDSSCDQRQD LPSEQNKGAS LDSSQCSPSP ASQEDLESEI SEDSDQEVDI EGDKSYFNAG

          Target Information

          HHEX is a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. Establishes anterior identity at two levels 1) acts early to enhance canonical WNT-signaling by repressing expression of TLE4 and 2) acts later to inhibit NODAL-signaling by directly targeting NODAL.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Hematopoietically-expressed homeobox protein HHEX; homeobox; Homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; Proline-rich homeodomain protein; proline-rich homeodomain-containing transcription factor

          View more View less

          Gene Aliases: HEX; HHEX; HMPH; HOX11L-PEN; PRH; PRHX

          View more View less

          UniProt ID: (Human) Q03014

          View more View less

          Entrez Gene ID: (Human) 3087

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding chromatin binding protein binding transcription factor binding eukaryotic initiation factor 4E binding DNA binding, bending TBP-class protein binding protein homodimerization activity sequence-specific DNA binding transcription regulatory region DNA binding repressing transcription factor binding HMG box domain binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter vasculogenesis in utero embryonic development myeloid leukocyte differentiation transcription, DNA-templated mRNA export from nucleus Notch signaling pathway endoderm development cell proliferation anterior/posterior pattern specification negative regulation of transcription by transcription factor localization negative regulation of transcription by competitive promoter binding Wnt signaling pathway negative regulation of angiogenesis poly(A)+ mRNA export from nucleus interkinetic nuclear migration cell differentiation positive regulation of Wnt signaling pathway B cell differentiation thyroid gland development negative regulation of vascular endothelial growth factor receptor signaling pathway pancreas development protein localization to nucleus embryonic heart tube development multicellular organism growth regulation of cell proliferation negative regulation of cyclin-dependent protein serine/threonine kinase activity negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter forebrain morphogenesis primary lung bud formation gall bladder development hepatic duct development hepatoblast differentiation hepatocyte differentiation DNA conformation change primitive streak formation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0