Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mfsd14a is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hippocampus abundant gene transcript 1; hippocampus abundant transcript 1; Hippocampus abundant transcript 1 protein; Major facilitator superfamily domain-containing 14A; Putative tetracycline transporter-like protein; tetracycline transporter-like protein
Gene Aliases: HIAT1; MFSD14A
UniProt ID: (Mouse) P70187, (Human) Q96MC6
Entrez Gene ID: (Mouse) 15247, (Human) 64645, (Rat) 103689987
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support