Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGQREMWRLM SRFNAFKRTN TILHHLRMSK HTDAAEEVLL GKKGCTGVIT LNRPKFLNAL TLNMIRQIYP QLKKWEQDPE TFLIIIKGAG GKAFCAGGDI RVISEAEKAK QKIAPVFFRE EYMLNNAVGS CQKPYVALIH GITMGGGVGL SVHGQFRVAT EKCLFAMPET AIGLFPDVGG GYFLPRLQGK LGYFLALTGF RLKGRDVYRA GIATHFVDSE KLAMLEEDLL ALKSPSKENI ASVLENYHTE SKIDRDKSFI LEEHMDKINS CFSANTVEEI IENLQQDGSS FALEQLKVIN KMSPTSLKIT LRQLMEGSSK TLQEVLTMEY RLSQACMRGH DFHEGVRAVL IDKDQSPKWK PADLKEVTEE DLNNHFKSLG SSDLKF
Beta-hydroxyisobutyryl-CoA hydrolase (EC 3.1.2.4) is responsible for the specific hydrolysis of HIBYL-CoA, a valine catabolite, as well as the hydrolysis of beta-hydroxypropionyl-CoA, an intermediate in a minor pathway of propionate metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-hydroxyisobutyryl-CoA hydrolase; 3-hydroxyisobutyryl-CoA hydrolase, mitochondrial; 3-hydroxyisobutyryl-Coenzyme A hydrolase; HIB-CoA hydrolase; HIBYL-COA-H; testicular tissue protein Li 86
Gene Aliases: 2610509I15Rik; AI648812; HIBCH; HIBYLCOAH
UniProt ID: (Human) Q6NVY1, (Rat) Q5XIE6, (Mouse) Q8QZS1
Entrez Gene ID: (Rabbit) 100342287, (Human) 26275, (Rat) 301384, (Mouse) 227095
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support