Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSETVPPAPA ASAAPEKPLA GKKAKKPAKA AAASKKKPAG PSVSELIVQA ASSSKERGGV SLAALKKALA AAGYDVEKNN SRIKLGIKSL VSKGTLVQTK GTGASGSFKL NKKASSVETK PGASKVATKT KATGASKKLK KATGASKKSV KTPKKAKKPA ATRKSSKNPK KPKTVKPKKV AKSPAKAKAV KPKAAKARVT KPKTAKPKKA APKKK
This locus is the ortholog of a mouse protein-coding gene that displays characteristics of a linker histone and is expressed in nuclei of late maturing spermatids. The human locus is expressed; however, the open reading frame has been disrupted by a frameshift so it is unlikely to encode a functional protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H1 antibody; H1 histone family, member 0; H1 histone family, member 1; H1 histone family, member 2; H1 histone family, member 5; H1(0); H1-0; H1.0; H1.0, H1(0), H1-0; H10; H1F3; H1FV; HIST1H1B antibody; HIST1H1D; Histone 1 antibody; histone 1, H1a; histone 1, H1b; histone 1, H1c; Histone H1'; Histone H1(0); Histone H1.0; Histone H1.1; Histone H1.2; Histone H1.3; Histone H1.5; Histone H1a; Histone H1b; Histone H1c; Histone H1d; Histone H1s-1; Histone H1s-2; Histone H1s-3
Gene Aliases: H1; H1-0; H1-1; H1-2; H1-5; H1.1; H1.2; H1.5; H10; H1A; H1B; H1C; H1F0; H1F1; H1F2; H1F5; H1FV; H1s-1; H1s-3; HIST1; HIST1H1A; HIST1H1B; HIST1H1C
UniProt ID: (Human) P07305, (Human) Q02539, (Human) P16401, (Human) P16403
Entrez Gene ID: (Human) 3005, (Human) 3024, (Human) 3009, (Human) 3006
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support