Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MSETVPPAPA ASAAPEKPLA GKKAKKPAKA AAASKKKPAG PSVSELIVQA ASSSKERGGV SLAALKKALA AAGYDVEKNN SRIKLGIKSL VSKGTLVQTK GTGASGSFKL NKKASSVETK PGASKVATKT KATGASKKLK KATGASKKSV KTPKKAKKPA ATRKSSKNPK KPKTVKPKKV AKSPAKAKAV KPKAAKARVT KPKTAKPKKA APKKK
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H1 histone family, member 1; histone 1, H1a; Histone H1.1; Histone H1a
Gene Aliases: H1-1; H1.1; H1A; H1F1; HIST1; HIST1H1A
UniProt ID: (Human) Q02539
Entrez Gene ID: (Human) 3024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support