Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MRPEDRMFHI RAVILRALSL AFLLSLRGAG AIKADHVSTY AAFVQTHRPT GEFMFEFDED EQFYVDLDKK ETVWHLEEFG RAFSFEAQGG LANIAILNNN LNTLIQRSNH TQAANDPPEV TVFPKEPVEL GQPNTLICHI DRFFPPVLNV TWLCNGEPVT EGVAESLFLP RTDYSFHKFH YLTFVPSAED VYDCRVEHWG LDQPLLKHWE AQEPIQMPET TETVLCALGL VLGLVGIIVG TVLIIKSLRS GHDPRAQGPL
HLA-DPA1 belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta (DPB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and the cytoplasmic tail. Within the DP molecule both the alpha chain and the beta chain contain the polymorphisms specifying the peptide binding specificities, resulting in up to 4 different molecules.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DP(W3); DP(W4); HLA class II histocompatibility antigen, DP alpha 1 chain; HLA-SB alpha chain; MHC class II antigen; MHC class II DP3-alpha; MHC class II DPA1; MHC class II HLA-DPA1 antigen; Primed lymphocyte test-1
Gene Aliases: DP(W3); DP(W4); HLA-DP1A; HLA-DPA1; HLADP; HLASB; PLT1
UniProt ID: (Human) Q5STP1
Entrez Gene ID: (Human) 3113
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support