Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HLA-DQ Monomorphic Antibodies

          Leinco Technologies

          HLA-DQ (MHC Class II) Monoclonal Antibody (1a3), Functional Grade, In Vivo PLATINUM™

          View all (2) HLA-DQ Monomorphic antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite HLA-DQ (MHC Class II) Monoclonal Antibody (1a3), Functional Grade, In Vivo PLATINUM™

          Product Details

          H462-1MG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1a3

          Immunogen

          Proprietary immunogen

          Conjugate

          Functional Grade Functional Grade Functional Grade

          Form

          Liquid

          Concentration

          ≥5.0 mg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.2-7.4

          Contains

          no preservative

          Storage conditions

          4°C short term, -80°C long term

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Specificity: Clone 1a3 recognizes a monomorphic epitope on human HLA-DQ1. It does not cross-react with HLA-DR or HLA-DP.

          Endotoxin Level: <0.5 EU/mg as determined by the LAL method. Purity: >95% by SDS Page; ≥98% monomer by analytical SEC.

          Product Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using only In vitro protein free cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUMTM antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile.

          Storage and Handling: Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80 C. Avoid Repeated Freeze Thaw Cycles.

          This antibody has been tested in the following applications: Western Blotting, Immunoprecipitation, Flow Cytometry, ELISA.

          Target Information

          MHC class II is constitutively expressed on human professional antigen-presenting cells (APCs), including macrophages/monocytes, dendritic cells (DCs), and B cells, and is induced on T cells upon activation. HLA-DQ consists of two transmembrane proteins, a 35 kDa alpha (heavy) chain and 29 kDa Beta (light) chain 3 encoded by the HLA-DQA1 and HLA-DQB1 genes, respectively, located in the HLA complex of chromosome 6. The N-terminal alpha-1 and Beta-1 domains form the antigen-binding groove, which binds 13-25 aa peptides derived from exogenous antigens. On APCs, MHC class II plays a critical role in the adaptive immune response by presenting phagocytosed antigens to helper CD4 T cells. The T cell receptor (TCR)/CD3 complex of CD4 T cells interacts with peptide-MHC class II, which induces CD4 T cell activation leading to the coordination and regulation of other effector cells. CD4 molecules also bind to MHC class II, which helps augment TCR signaling. It has also been demonstrated that MHC class II express on activated T cells are capable of antigen presentation and can transduce signals into T cells, enhancing T cell proliferation and activity. Specific alleles of HLA-DQ are associated with autoimmune diseases, including celiac disease and type 1 diabetes, and graft-versus-host disease.

          HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DC-1 alpha chain; DC-alpha; HLA class II antigen; HLA class II histocompatibility antigen, DQ alpha 1 chain; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA class II histocompatibility antigen, DQ(W3) alpha chain; HLA-DCA; leucocyte antigen DQA1; leukocyte antigen alpha chain; lymphocyte antigen; MHC class II antigen; MHC class II antigen DQB1; MHC class II antigen HLA-DQ-beta-1; MHC class II DQ beta chain; MHC class II DQA1; MHC class II HLA-D alpha glycoprotein; MHC class II HLA-DQ beta glycoprotein; MHC class II HLA-DQ-alpha-1; MHC class II surface glycoprotein; MHC class2 antigen; MHC DQ beta; MHC HLA-DQ alpha

          View more View less

          Gene Aliases: CD; CELIAC1; DQ-A1; GSE; HLA-DQA; HLA-DQA1; HLA-DQB; HLA-DQB1; HLA-DRB1; IDDM1

          View more View less

          UniProt ID: (Human) P01909, (Human) P01920

          View more View less

          Entrez Gene ID: (Human) 3117, (Human) 3119

          View more View less

          Function(s)
          MHC class II receptor activity peptide antigen binding
          Process(es)
          immunoglobulin production involved in immunoglobulin mediated immune response humoral immune response mediated by circulating immunoglobulin immune response antigen processing and presentation of exogenous peptide antigen via MHC class II T cell costimulation T cell receptor signaling pathway interferon-gamma-mediated signaling pathway
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0