Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LEDRVRGPK GPEEKKDCEV VMMIGLPGAG KTTWVTKHAA ENPGKYNILG TNTIMDKMMV AGFKKQMADT GKLNTLLQRA PQCLGKFIEI AARKKRNFIL DQTNVSAAAQ RRKMCLFAGF QRKAVVVCPK DEDYKQRTQK KAEVEGKDLP EHAVLKMKGN FTLPEVAECF DEITYVELQK EEAQKLLEQY KEESKKALPP EKKQNTGSKK SNKNKSGKNQ FNRGGGHRGR GGFNMRGGNF RGGAPGNRGG YNRRGNMPQR GGGGGGSGGI GYPYPRAPVF PGRGSYSNRG NYNRGGMPNR GNYNQNFRGR GNNRGYKNQS QGYNQWQQGQ FWGQKPWSQH YHQGYY (462-806 aa encoded by BC003367)
The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ30202; FLJ37978; GRIP120; Heterogeneous nuclear ribonucleoprotein U; heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A); heterogenous nuclear ribonucleoprotein U; hnRNP U; HNRNPU antisense RNA 1; NCRNA00201; nuclear matrix protein sp120; Nuclear p120 ribonucleoprotein; p120; p120 nuclear protein; pp120; SAF-A; scaffold attachment factor A; Scaffold-attachment factor A; SP120; system N1 Na+ and H+-coupled glutamine transporter; transporter protein; transporter protein; system N1 Na+ and H+-coupled glutamine transporter; U211
Gene Aliases: AA408410; AI256620; AL024194; AL024437; AW557595; C1orf199; C86794; hnRNP U; HNRNPU; HNRNPU-AS1; HNRPU; rCG_20317; SAF-A; SAFA; SN1; Sp120; U21.1
UniProt ID: (Human) Q00839, (Rat) Q6IMY8, (Mouse) Q8VEK3
Entrez Gene ID: (Human) 3192, (Rat) 117280, (Mouse) 51810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support