Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HO-1 Antibodies

          Proteintech

          HO-1/HMOX1 Monoclonal Antibody (2D10A5)

          93 References
          View all (38) HO-1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite HO-1/HMOX1 Monoclonal Antibody (2D10A5)

          • Antibody Testing Data (11)
          HO-1/HMOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          HO-1/HMOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HO-1/HMOX1 Antibody (66743-1-IG) in IHC (P)

          Immunohistochemistry of paraffin-embedded human renal cell carcinoma tissue slide using 66743-1-Ig (HO-1/HMOX1 antibody) at dilution of 1:1000 (under 40x lens) heat mediated antigen retrieved with Tris-EDTA buffer (pH 9). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HO-1/HMOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HO-1/HMOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HO-1/HMOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HO-1/HMOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HO-1/HMOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HO-1/HMOX1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HO-1/HMOX1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          HO-1/HMOX1 Antibody in Western Blot (WB)
          HO-1/HMOX1 Antibody in Western Blot (WB)
          HO-1/HMOX1 Antibody in Western Blot (WB)
          HO-1/HMOX1 Antibody in Western Blot (WB)

          Product Details

          66743-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:6,000
          View 79 publications 79 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:500-1:2,000
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          -
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rabbit, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2D10A5

          Immunogen

          HO-1/HMOX1 Fusion Protein Ag21296 (1-288 aa encoded by BC001491)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MERPQPDSM PQDLSEALKE ATKEVHTQAE NAEFMRNFQK GQVTRDGFKL VMASLYHIYV ALEEEIERNK ESPVFAPVYF PEELHRKAAL EQDLAFWYGP RWQEVIPYTP AMQHYVKRLH EVGRTEPELL VAHAYTRYLG DLSGGQVLKK IAQKALDLPS SGEGLAFFTF PNIASATKFK QLYRSRMNSL EMTPAVRQRV IEEAKTAFLL NIQLFEELQE LLTHDTKDQS PSRAPGLRQR ASNKVQDSAP VETPRGKPPL NTRSQAPLLR WVLTLSFLVA TVAVGLYAM (1-288 aa encoded by BC001491 )

          Target Information

          Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; Heme oxygenase 1; hemoxygenase; HO-1; HSP32; P32 protein

          View more View less

          Gene Aliases: bK286B10; D8Wsu38e; Hemox; Heox; HEOXG; Hmox; HMOX1; HMOX1D; HO; HO-1; HO1; HSP32

          View more View less

          UniProt ID: (Human) P09601, (Pig) P32394, (Rat) P06762, (Mouse) P14901

          View more View less

          Entrez Gene ID: (Human) 3162, (Pig) 445512, (Rabbit) 100008919, (Rat) 24451, (Mouse) 15368

          View more View less

          Function(s)
          heme oxygenase (decyclizing) activity phospholipase D activity signal transducer activity protein binding enzyme binding heme binding protein homodimerization activity metal ion binding oxidoreductase activity
          Process(es)
          angiogenesis endothelial cell proliferation wound healing involved in inflammatory response negative regulation of leukocyte migration heme oxidation cellular iron ion homeostasis response to oxidative stress small GTPase mediated signal transduction excretion regulation of blood pressure cell death intrinsic apoptotic signaling pathway in response to DNA damage negative regulation of muscle cell apoptotic process smooth muscle hyperplasia cellular response to nutrient negative regulation of mast cell cytokine production erythrocyte homeostasis low-density lipoprotein particle clearance regulation of transcription from RNA polymerase II promoter in response to iron cellular response to heat response to nicotine intracellular signal transduction heme catabolic process response to hydrogen peroxide positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of mast cell degranulation negative regulation of DNA binding negative regulation of sequence-specific DNA binding transcription factor activity negative regulation of neuron apoptotic process regulation of transcription from RNA polymerase II promoter in response to oxidative stress response to estrogen positive regulation of chemokine biosynthetic process regulation of angiogenesis positive regulation of angiogenesis positive regulation of vasodilation positive regulation of smooth muscle cell proliferation negative regulation of smooth muscle cell proliferation regulation of sequence-specific DNA binding transcription factor activity protein homooligomerization iron ion homeostasis cellular response to arsenic-containing substance cellular response to cadmium ion cellular response to hypoxia negative regulation of extrinsic apoptotic signaling pathway via death domain receptors response to hypoxia phospholipid metabolic process negative regulation of cell proliferation heme metabolic process apoptotic process oxidation-reduction process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0