Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSTAREQPIF STRAHVFQID PATKRNWIPA GKHALTVSYF YDATRNVYRI ISIGGAKAII NSTVTPNMTF TKTSQKFGQW ADSRANTVYG LGFASEQHLT QFAEKFQEVK EAARLAREKS QDGGELTSPA LGLASHQVPP SPLVSANGPG EEKLFRSQSA DAPGPTERER LKKMLSEGSV GEVQWEAEFF ALQDSNNKLA GALREANAAA AQWRQQLEAQ RAEAERLRQR VAELEAQAAS EVTPTGEKEG LGQGQSLEQL EALVQTKDQE IQTLKSQTGG PREALEAAER EETQQKVQDL ETRNAELEHQ LRAMERSLEE ARAERERARA EVGRAAQLLD VRLFELSELR EGLARLAEAA P
This gene encodes a member of the HOMER family of postsynaptic density scaffolding proteins that share a similar domain structure consisting of an N-terminal Enabled/vasodilator-stimulated phosphoprotein homology 1 domain which mediates protein-protein interactions, and a carboxy-terminal coiled-coil domain and two leucine zipper motifs that are involved in self-oligomerization. The encoded protein binds numerous other proteins including group I metabotropic glutamate receptors, inositol 1,4,5-trisphosphate receptors and amyloid precursor proteins and has been implicated in diverse biological functions such as neuronal signaling, T-cell activation and trafficking of amyloid beta peptides. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homer homolog 3; Homer protein homolog 3; Homer, neuronal immediate early gene, 3; Homer-3
Gene Aliases: HOMER-3; HOMER3; VESL3
UniProt ID: (Human) Q9NSC5
Entrez Gene ID: (Human) 9454
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support