Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total HOXA11/D11.
HOX11, also called homeobox11 or Tcl-3 is located at the 10q24 T-Cell translocation breakpoint region. It encodes a homeobox-domain containing protein, HOX11, containing a glycine and proline-rich amino terminus. HOX11 is required for maintenance of the developing spleen and cell survival. The HOX11protein binds to a specific DNA sequence and localizes to the cell nucleus. It transactivates transcription of a reporter gene linked to a cis-regulatory element, acting as a positive transcription activator. The HOX11 gene is highly expressed in T-ALLs as a result of a t (7:10) (q35;q24) or t(10:14)(q24;qll) translocation. The homeobox gene is deregulated upon translocation into the T-cell receptor locus and activates a specific subset of tumor-associated target genes. HOX 11 gene translocation has been attributed to T-cell leukemia and lymphoid neoplasias.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box 1I; homeo box 4F; homeo box A11; homeo box D11; Homeobox protein Hox-1.9; Homeobox protein Hox-1I; Homeobox protein Hox-4.6; Homeobox protein Hox-4F; Homeobox protein Hox-5.5; Homeobox protein Hox-A11; Homeobox protein Hox-D11; homeobox protein HOXA11; Hox-4.6, mouse, homolog of; HOX11
Gene Aliases: E230017H14Rik; Hox-1.9; Hox-4.6; Hox-5.4; Hox-5.5; HOX1; HOX1I; HOX4; HOX4F; Hoxa-11; HOXA11; Hoxd-11; HOXD11; RUSAT1
UniProt ID: (Human) P31270, (Human) P31277, (Mouse) P31311, (Mouse) P23813
Entrez Gene ID: (Human) 3207, (Human) 3237, (Mouse) 15396, (Mouse) 15431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support