Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: RTAYTRQQVL ELEKEFHYNR YLTRRRRIEI AHSLCLSERQ IKIWFQNRRM KWKKDHRLPN TKVRSAPPAG AAPSTLSAAT PGTSEDHSQS ATPPEQQRAE DITRL
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box 3E; Homeobox protein CP19; Homeobox protein Hox-3E; Homeobox protein Hox-C4; Hox-35
Gene Aliases: cp19; HOX3; HOX3E; HOXC4
UniProt ID: (Human) P09017
Entrez Gene ID: (Human) 3221
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support