Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: PFYSPQENVV FSSSRGPYDY GSNSFYQEKD MLSNCRQNTL GHNTQTSIAQ DFSSEQGRTA PQDQKASIQI YPWMQRMNSH SGVGYGADRR
This protein belongs to the homeobox family, members of which encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene, HOXC6, is one of several HOXC genes located in a cluster on chromosome 12. Three genes, HOXC5, HOXC4 and HOXC6, share a 5' non-coding exon.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box 3C; homeo box C8 protein; Homeobox protein CP25; Homeobox protein HHO.C8; Homeobox protein Hox-3C; Homeobox protein Hox-C6
Gene Aliases: CP25; HHO.C8; HOX3; HOX3C; HOXC6
UniProt ID: (Human) P09630
Entrez Gene ID: (Human) 3223
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support