Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VLARTDWTV EDGLQKYERG LIFYINHSLY ENLDEELNEE LAAKVVQMFY VAEPKQVPHI LCSPSMKNIN PLTAMSYLRK LDTSGFSSIL VTLTKAAVAL KMGDLDMHRN EMKSHSEMKL VCGFILEPRL LIQQRKGQIV PTELALHLKE TQPGLLVASV LGLQKNNKIG IEEADSFFKV LCAKDEDTIP QLLVDFWEAQ LVACLPDVVL QELFFKLTSQ YIWRLSKRQP PDTTPLRTSE DLINACSHYG LIYPWVHVVI SSDSLADKNY TEDLSKLQSL ICGPSFDIAS IIPFLEPLSE DTIAGLSVHV LCRTRLKEYE QCIDILLERC PEAVIPYANH ELKEENRTLW WKKLLPELCQ RIKCGGEKYQ LYLSSLKETL SIVAVELELK DFMNVLPEDG TATFFLPYLL YCSRKKPLT (441-858 aa encoded by BC022062)
This gene encodes a protein containing a potential clathrin-binding motif, consensus dileucine signals, and tyrosine-based sorting signals for targeting to vesicles of lysosomal lineage. The encoded protein may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 3. Alternate splice variants exist, but their full length sequence has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BLOC-2 complex member HPS3; cocoa; DKFZp686F0413; FLJ22704; Hermansky-Pudlak syndrome 3 protein
Gene Aliases: BLOC2S1; HPS3; SUTAL
UniProt ID: (Human) Q969F9
Entrez Gene ID: (Human) 84343
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support