Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • AGFG1 Antibodies

          Abnova

          HRB Polyclonal Antibody, MaxPab™

          View all (9) AGFG1 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite HRB Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          HRB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HRB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HRB Antibody (H00003267-B01P) in WB

          Western Blot analysis of AGFG1 expression in transfected 293T cell line by AGFG1 MaxPab polyclonal antibody. Lane 1: HRB transfected lysate(61.82 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HRB Antibody in Western Blot (WB)

          Product Details

          H00003267-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          HRB (NP_004495, 1 a.a. approximately 562 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAASAKRKQE EKHLKMLRDM TGLPHNRKCF DCDQRGPTYV NMTVGSFVCT SCSGSLRGLN PPHRVKSISM TTFTQQEIEF LQKHGNEVCK QIWLGLFDDR SSAIPDFRDP QKVKEFLQEK YEKKRWYVPP EQAKVVASVH ASISGSSASS TSSTPEVKPL KSLLGDSAPT LHLNKGTPSQ SPVVGRSQGQ QQEKKQFDLL SDLGSDIFAA PAPQSTATAN FANFAHFNSH AAQNSANADF ANFDAFGQSS GSSNFGGFPT ASHSPFQPQT TGGSAASVNA NFAHFDNFPK SSSADFGTFN TSQSHQTASA VSKVSTNKAG LQTADKYAAL ANLDNIFSAG QGGDQGSGFG TTGKAPVGSV VSVPSQSSAS SDKYAALAEL DSVFSSAATS SNAYTSTSNA SSNVFGTVPV VASAQTQPAS SSVPAPFGAT PSTNPFVAAA GPSVASSTNP FQTNARGATA ATFGTASMSM PTGFGTPAPY SLPTSFSGSF QQPAFPAQAA FPQQTAFSQQ PNGAGFAAFG QTKPVVTPFG QVAAAGVSSN PFMTGAPTGQ FPTGSSSTNP FL

          Target Information

          HRB is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. HRB binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. This encoded protein binds the Rev activation domain when Rev is assembled onto its RNA target and can significantly enhance Rev activity when overexpressed. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Arf-GAP domain and FG repeat-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; HIV-1 Rev binding protein; HIV-1 Rev-binding protein; hRIP, Rev interacting protein; MGC116938; MGC116940; Nucleoporin-like protein RIP; Rab, Rev/Rex activation domain-binding protein; Rev-interacting protein; Rev/Rex activation domain-binding protein

          View more View less

          Gene Aliases: AGFG1; HRB; RAB; RIP

          View more View less

          UniProt ID: (Human) P52594

          View more View less

          Entrez Gene ID: (Human) 3267

          View more View less

          Function(s)
          DNA binding RNA binding GTPase activator activity protein binding metal ion binding
          Process(es)
          acrosome assembly mRNA export from nucleus multicellular organism development spermatid nucleus differentiation positive regulation of GTPase activity intermediate filament organization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0