Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKALIAALLL ITLQYSCAVS PTDCSAVEPE AEKALDLINK RRRDGYLFQL LRIADAHLDR VENTTVYYLV LDVQESDCSV LSRKYWNDCE PPDSRRPSEI VIGQCKVIAT RHSHESQDLR VIDFNCTTSS VSSALANTKD SPVLIDFFED TERYRKQANK ALEKYKEEND DFASFRVDRI ERVARVRGGE GTGYFVDFSV RNCPRHHFPR HPNVFGFCRA DLFYDVEALD LESPKNLVIN CEVFDPQEHE NINGVPPHLG HPFHWGGHER SSTTKPPFKP HGSRDHHHPH KPHEHGPPPP PDERDHSHGP PLPQGPPPLL PMSCSSCQHA TFGTNGAQRH SHNNNSSDLH PHKHHSHEQH PHGHHPHAHH PHEHDTHRQH PHGHHPHGHH PHGHHPHGHH PHGHHPHCHD FQDYGPCDPP PHNQGHCCHG HGPPPGHLRR RGPGKGPRPF HCRQIGSVYR LPPLRKGEVL PLPEANFPSF PLPHHKHPLK PDNQPFPQSV SESCPGKFKS GFPQVSMFFT HTFPK
HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp779H1622; Histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; HPRG
Gene Aliases: HPRG; HRG; HRGP; THPH11
UniProt ID: (Human) P04196
Entrez Gene ID: (Human) 3273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support