Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: SATKTFVDFF SQCLHEEYRS KGVFVQSVLP YFVATKLAKI RKPTLDKPSP ETFVKSAIKT VGLQSRTNG
The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17-beta-HSD 12; 17-beta-hydroxysteroid dehydrogenase 12; 17beta-HSD type 12; 3-ketoacyl-CoA reductase; Estradiol 17-beta-dehydrogenase 12; hydroxysteroid (17-beta) dehydrogenase 12; KAR; keratoadhesin; keratonectin; KIK-I; Short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; smooth muscle-specific 17 beta-hydroxysteroid dehydrogenase type 3; steroid dehydrogenase; steroid dehydrogenase homolog; Very-long-chain 3-oxoacyl-CoA reductase
Gene Aliases: 2610510O05Rik; AI172963; HSD17B12; KAR; KIK-I; Kik1; SDR12C1
UniProt ID: (Human) Q53GQ0, (Rat) Q6P7R8, (Mouse) O70503
Entrez Gene ID: (Human) 51144, (Rat) 84013, (Mouse) 56348
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support