Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
1 µg/mL of MA5-45094 was sufficient for detection of HSP70/HSC70 in 20 µg of heat shocked HeLa cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 72 (HSP) and approximately 73kDa (HSC).
HSPA4 (Heat Shock Protein Family A (Hsp70) Member 4) is a Protein Coding gene. Diseases associated with HSPA4 include Chagas Disease and Babesiosis. Among its related pathways are Regulation of activated PAK-2p34 by proteasome mediated degradation and Cellular responses to stimuli. An important paralog of this gene is HSPH1. HSPA8 (Heat Shock Protein Family A (Hsp70) Member 8) is a Protein Coding gene. Diseases associated with HSPA8 include Auditory System Disease and Borna Disease. Among its related pathways are Processing of Capped Intron-Containing Pre-mRNA and Selective autophagy. Gene Ontology (GO) annotations related to this gene include RNA binding and ubiquitin protein ligase binding. An important paralog of this gene is HSPA2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: constitutive heat shock protein 70; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 5a; epididymis secretory sperm binding protein Li 72p; Heat shock 70 kDa protein 1; Heat shock 70 kDa protein 4; Heat shock 70 kDa protein 8; Heat shock 70-related protein APG-2; heat shock 70kd protein 10; heat shock 70kd protein 10 (HSC71); heat shock 70kD protein 4; heat shock 70kD protein 8; heat shock 70kDa protein 1-like; heat shock 70kDa protein 4; heat shock 70kDa protein 4-like; heat shock 70kDa protein 8; heat shock 70kDa protein 8-like; heat shock cognate 70; heat shock cognate 71 kD protein; Heat shock cognate 71 kDa protein; heat shock cognate hsc73; heat shock cognate protein 54; Heat shock cognate protein 70; heat shock protein 8; heat shock protein A8; heat shock protein Apg-2; heat shock protein cognate 70; Heat shock protein family A member 8; Heat shock protein family H member 2; heat shock protein, 110 kDa; Hsc70 ATPase; hsp70 RY; HSP70RY; HSPA4/HSPA8; Ischemia responsive 94 kDa protein; LAP-1; Lipopolysaccharide-associated protein 1; LPS-associated protein 1; N-myristoyltransferase inhibitor protein 71
Gene Aliases: 2410008N15Rik; 70kDa; AI317151; APG-2; APG2; BOS_14437; BOS_8056; HEL-33; HEL-S-5a; HEL-S-72p; HS24/P52; HSC54; HSC70; hsc70.I; HSC71; Hsc73; Hsp110; HSP70; Hsp70RY; HSP71; HSP73; HSPA1; HSPA10; hspa1l; HSPA4; HSPA8; HSPH2; I79_004332; I79_022974; Irp94; LAP-1; LAP1; mKIAA4025; NIP71; RY; XELAEV_18037286mg
UniProt ID: (Dog) Q7YQC6, (Dog) Q2TFN9, (Human) P34932, (Bovine) P19120, (Human) P11142, (Chicken) O73885, (Rat) O88600, (Chinese hamster) P19378, (Rat) P63018, (Mouse) P63017
Entrez Gene ID: (Dog) 403612, (Bovine) 536558, (Dog) 474680, (Sheep) 101122782, (Human) 3308, (Bovine) 281831, (Dog) 479406, (Sheep) 101112844, (Human) 3312, (Chicken) 395853, (Pig) 100511890, (Chinese hamster) 100764628, (Mouse) 15525, (Rat) 266759, (Chinese hamster) 100689472, (Rat) 24468, (Mouse) 15481, (Rabbit) 100338049, (Rabbit) 100348873, (Xenopus) 379760
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support